Learn More
WTW™ 4-Electrode Flow-through Conductivity Cell
GREENER_CHOICE
Product Code. 10089760
Change view
Click to view available options
For Use With (Equipment):
pH Electrode
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10089760 pH Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10089760 Supplier WTW™ Supplier No. 301254
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Flow measuring cells by WTW for medium and high conductivity are suitable for extensive measuring series in the lab but also for the maintenance and measurement of cleaning solutions in special systems.
TRUSTED_SUSTAINABILITY
Specifications
Description Conductivity cells
For Use With (Equipment) pH Electrode
Includes Built-in temperature sensor