missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylphosphinyl)benzenamine Hydrochloride, TRC

Product Code. 30380069
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380069 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380069 Supplier Toronto Research Chemicals Supplier No. TRCD477610250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Dimethylphosphinyl)benzenamine Hydrochloride
CAS 479353-60-1
Molecular Formula C8H12NOP • HCl
Quantity 250 mg
Synonym 4-(Dimethylphosphoryl)benzenamine Hydrochloride, 4-(Dimethylphosphinoyl)aniline Hydrochloride, (4-Aminophenyl)dimethylphosphine Oxide Hydrochloride
InChI Formula InChI=1S/C8H12NOP.ClH/c1-11(2,10)8-5-3-7(9)4-6-8;/h3-6H,9H2,1-2H3;1H
SMILES NC1=CC=C(P(C)(C)=O)C=C1.Cl
IUPAC Name (4-aminophenyl)dimethylphosphine oxide hydrogen chloride.
Molecular Weight (g/mol) 205.62
Formula Weight 205.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.