missing translation for 'onlineSavingsMsg'
Learn More

4'-Dimethylaminophenyl Acetylene, TRC

Product Code. 30378691
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378691 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378691 Supplier Toronto Research Chemicals Supplier No. TRCD456710250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Dimethylaminophenyl Acetylene
CAS 17573-94-3
Molecular Formula C10H11N
Quantity 250 mg
Synonym 4-Ethynyl-N,N-dimethylbenzenamine, (4-Ethynylphenyl)dimethylamine, 1-(Dimethylamino)-4-ethynylbenzene, 1-Ethynyl-4-(dimethylamino)benzene, 4-(Dimethylamino)phenylethyne, 4-Dimethylaminophenylacetylene, 4-Ethynyl-N,N-dimethylaminobenzene, 4-Ethynyl-N,N-dimethylaniline, 4-Ethynylphenyl-N,N-dimethylamine, N,N-Dimethyl-4-ethynylaniline, N,N-Methyl-4-ethynylaniline, N-(4-Ethynylphenyl)-N,N-dimethylamine, [4-(N,N-Dimethylamino)phenyl]acetylene, [p-(Dimethylamino)phenyl]acetylene, p-Ethynyl-N,N-dimethylaniline
InChI Formula InChI=1S/C10H11N/c1-4-9-5-7-10(8-6-9)11(2)3/h1,5-8H,2-3H3
SMILES CN(C)c1ccc(cc1)C#C
IUPAC Name 4-ethynyl-N,N-dimethylaniline
Molecular Weight (g/mol) 145.2
Formula Weight 145.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.