Learn More
4-Dimethylamino-4'-methylazobenzene, TRC
Product Code. 30179952
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179952 100 mg 100mg
30179891 500 mg 500mg
30179839 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179952 Supplier Toronto Research Chemicals Supplier No. TRCD826565100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3010-57-9
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.32
SMILES CN(C)c1ccc(cc1)N=Nc2ccc(C)cc2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H17N3/c1-12-4-6-13(7-5-12)16-17-14-8-10-15(11-9-14)18(2)3/h4-11H,1-3H3
Formula Weight 239.1422
Chemical Name or Material 4-Dimethylamino-4'-methylazobenzene