Learn More
4-Desmethoxypropoxyl-4-methoxy S-Deoxo Rabeprazole, TRC
Product Code. 30008411
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008411 25 mg 25mg
30004413 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008411 Supplier Toronto Research Chemicals Supplier No. TRCD29061525MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102804-82-0
Molecular Formula C15 H15 N3 O S
Molecular Weight (g/mol) 285.36
Synonym 2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]thio]-1H-benzimidazole, Rabeprazole Sodium Imp G (EP)
IUPAC Name 2-[(4-methoxy-3-methylpyridin-2-yl)methylsulfanyl]-1H-benzimidazole
SMILES COc1ccnc(CSc2nc3ccccc3[nH]2)c1C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C15H15N3OS/c1-10-13(16-8-7-14(10)19-2)9-20-15-17-11-5-3-4-6-12(11)18-15/h3-8H,9H2,1-2H3,(H,17,18)
Formula Weight 285.0936
Percent Purity >95%
Chemical Name or Material 4-Desmethoxypropoxyl-4-methoxy S-Deoxo Rabeprazole
Grade HPLC