missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzoyl chloride, 98%

Product Code. p-7027520
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11495757

Brand: Thermo Scientific Alfa Aesar B24333.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6068-72-0 | C8H4ClNO | 165.58 g/mol

It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.

Solubility
Reacts with water.

Notes
Store under dry inert gas. Keep away from moisture, oxidizing agents, acids, bases, active metals, amines and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6068-72-0
Molecular Formula C8H4ClNO
Molecular Weight (g/mol) 165.58
MDL Number MFCD00001822
InChI Key USEDMAWWQDFMFY-UHFFFAOYSA-N
Synonym 4-cyanobenzoylchloride, 4-cyano-benzoyl chloride, p-cyanobenzoyl chloride, unii-2577fde7a8, benzoyl chloride, 4-cyano, 4cyanobenzoyl chloride, 4-cyano-benzoylchloride, 4-cyanobenzoyl-chloride, 4-cyanpbenzoyl chloride, 4-cyano benzoylchloride
PubChem CID 80172
IUPAC Name 4-cyanobenzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 68°C to 70°C
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 386729
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 165.58
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzoyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.