Learn More
4-Chromanol, 97%
Product Code. 11412777
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11412777
Brand: Thermo Scientific Alfa Aesar A14940.03
Request Bulk or Custom Format
CAS: 1481-93-2 | C9H10O2 | 150.177 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1481-93-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00006849
InChI Key MGSHXMOLUWTMGP-UHFFFAOYSA-N
PubChem CID 92890
IUPAC Name 3,4-dihydro-2H-chromen-4-ol
SMILES C1COC2=CC=CC=C2C1O
Specifications
Melting Point 38°C to 42°C
Boiling Point 79°C to 81°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 130800
Synonym 4-chromanol, chroman-4-ol, 3,4-dihydro-2h-1-benzopyran-4-ol, 4-hydroxychroman, 2h-1-benzopyran-4-ol, 3,4-dihydro, acmc-20mukj, 3'-carboxy-alpha-chromanol, 2h-1-benzopyran-4-ol,3,4-dihydro
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 4-Chromanol