Learn More
4-Chlorotoluene-d7, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Chlorotoluene-D7 |
| CAS | 84344-06-9 |
| Molecular Formula | C7 D7 Cl |
| Quantity | 0.1 g |
| Synonym | Benzene-1,2,4,5-d4, 3-chloro-6-(methyl-d3)- (9 CI), 3-Chloro-6-(methyl-d3)benzene-1,2,4,5-d4, 1-Chloro-4-(trideuteriomethyl)-2,3,5,6-tetradeuteriobenzene, 4-Chlorotoluene-d7, 4-Chlorotoluene-D7, 1-Chloro-4-(methyl-d3)benzene-2,3,5,6-d4 |
| InChI Formula | InChI=1 S/C7H7Cl/c1-6-2-4-7(8)5-3-6/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Cl)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-chloro-2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 133.6266 |
| Formula Weight | 133.0676 g/mol |
| Show More |