missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorothiobenzamide-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Chlorothiobenzamide-2,3,5,6-D4 |
| CAS | 1219802-58-0 |
| Molecular Formula | C7 D4 H2 Cl N S |
| Quantity | 0.1 g |
| Synonym | 4-Chlorothiobenzamide-2,3,5,6-D4, 4-Chlorobenzene-2,3,5,6-d4-carbothioamide, 4-Chlorobenzothiamide-d4 |
| InChI Formula | InChI=1 S/C7H6ClNS/c8-6-3-1-5(2-4-6)7(9)10/h1-4 H,(H2,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C(=S)N)c([2 H])c([2 H])c1Cl |
| IUPAC Name | 4-chloro-2,3,5,6-tetradeuteriobenzenecarbothioamide |
| Molecular Weight (g/mol) | 175.6719 |
| Formula Weight | 175.0161 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction