Learn More
4-Chlorophenol-d4, TRC
Product Code. 30364632
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364632 250 mg 250mg
30356555 100 mg 100mg
30360887 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30364632 Supplier Toronto Research Chemicals Supplier No. TRCC375042250MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 285132-91-4
Molecular Formula C6 2H4 H Cl O
Molecular Weight (g/mol) 132.58
Synonym Phen-2,3,5,6-d4-ol, 4-chloro- (9CI), 4-Chlorophenol-D4, p-Chlorophenol-d4, 4-Chlorophenol D4 (phenyl D4)
IUPAC Name 4-chloro-2,3,5,6-tetradeuteriophenol
SMILES [2H]c1c([2H])c(Cl)c([2H])c([2H])c1O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H/i1D,2D,3D,4D
Formula Weight 132.028
Chemical Name or Material 4-Chlorophenol ring-d4