Learn More
4-Chloromethyl-2-methylthiazole hydrochloride, 98%, Thermo Scientific™
Product Code. 10712443 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10712443 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10712443 Supplier Thermo Scientific Acros Supplier No. 355640010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 77470-53-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 161°C to 163°C
Color Brown or Cream to Gray-Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C5H7Cl2NS
MDL Number MFCD00067725
Quantity 1 g
Synonym 4-chloromethyl-2-methylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-methylthiazole hcl, 4-chloromethyl-2-methyl thiazole hydrochloride, 2-methyl-4-chloromethyl thiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole, chloride, 2-methyl-4-chloromethylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hcl
InChI Key YGKDISJLDVGNOR-UHFFFAOYSA-N
SMILES Cl.CC1=NC(CCl)=CS1
IUPAC Name 4-(chloromethyl)-2-methyl-1,3-thiazole;hydrochloride
Molecular Weight (g/mol) 184.08
PubChem CID 2734203
Formula Weight 184.09
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloromethyl-2-methylthiazole hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloromethyl-2-methylthiazole hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.