Learn More
4-Chloroisatin, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloroisatin |
| CAS | 6344-05-4 |
| Molecular Formula | C8H4ClNO2 |
| Quantity | 1 g |
| Synonym | 4-Chloro-1H-indole-2,3-dione, 4-Chloroindoline-2,3-dione, 4-Chloroisatin, NSC 49794 |
| InChI Formula | InChI=1S/C8H4ClNO2/c9-4-2-1-3-5-6(4)7(11)8(12)10-5/h1-3H,(H,10,11,12) |
| SMILES | Clc1cccc2NC(=O)C(=O)c12 |
| IUPAC Name | 4-chloro-1H-indole-2,3-dione |
| Molecular Weight (g/mol) | 181.58 |
| Formula Weight | 180.9931 |