missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyl Chloroformate, TRC

Product Code. 30359645
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30359645 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359645 Supplier Toronto Research Chemicals Supplier No. TRCC29286510ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobutyl Chloroformate
CAS 37693-18-8
Molecular Formula C5H8Cl2O2
Quantity 10 mL
Synonym 4-Chlorobutyl Ester Carbonochloridic Acid, 4-Chlorobutyl Carbonochloridate
InChI Formula InChI=1S/C5H8Cl2O2/c6-3-1-2-4-9-5(7)8/h1-4H2
SMILES ClCCCCOC(=O)Cl
IUPAC Name 4-chlorobutyl carbonochloridate
Molecular Weight (g/mol) 171.02
Formula Weight 169.9901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.