missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzamide, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 4-Chlorobenzamide |
| CAS | 619-56-7 |
| Molecular Formula | C7 H6 Cl N O |
| Quantity | 50 mg |
| Synonym | Benzamide, 4-chloro-, Benzamide, p-chloro- (6CI,7CI,8CI), 4-Chlorobenzamide, NSC 406894, NSC 74687, p-Chlorobenzamide, p-Chlorobenzoic acid amide |
| InChI Formula | InChI=1S/C7H6ClNO/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H2,9,10) |
| SMILES | NC(=O)c1ccc(Cl)cc1 |
| IUPAC Name | 4-chlorobenzamide |
| Molecular Weight (g/mol) | 155.58 |
| Formula Weight | 155.0138 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction