Learn More
4-Chloroaniline, 98%
Product Code. p-7020804
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
Product Code. 11495313
Brand: Thermo Scientific Alfa Aesar A12879.30
Request Bulk or Custom Format
CAS: 106-47-8 | C6H6ClN | 127.571 g/mol
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.
Solubility
Soluble in water (2.6 g/Liter at 20°C ).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, chloroformates and nitrous acid.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-47-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00007835
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
Synonym p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
PubChem CID 7812
ChEBI CHEBI:20331
IUPAC Name 4-chloroaniline
SMILES C1=CC(=CC=C1N)Cl
Specifications
Melting Point 67°C to 72°C
Density 1.43
Boiling Point 232°C
Quantity 250 g
UN Number UN2018
Beilstein 471359
Merck Index 14,2118
Solubility Information Soluble in water (2.6g/Liter at 20°C ).
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 4-Chloroaniline