Learn More
4-Chloro-7-nitro-2,1,3-benzothiadiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-7-nitro-2,1,3-benzothiadiazole |
| CAS | 2207-29-6 |
| Molecular Formula | C6H2N3O2SCl |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C6H2ClN3O2S/c7-3-1-2-4(10(11)12)6-5(3)8-13-9-6/h1-2H |
| SMILES | [O-][N+](=O)c1ccc(Cl)c2nsnc12 |
| IUPAC Name | 4-chloro-7-nitro-2,1,3-benzothiadiazole |
| Molecular Weight (g/mol) | 215.62 |
| Formula Weight | 214.9556 |