Learn More
4-Chloro-6-fluoro-2-methylquinazoline, TRC
Product Code. 30363382
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363382 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30363382 Supplier Toronto Research Chemicals Supplier No. TRCC5850481G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Chloro-6-fluoro-2-methylquinazoline
CAS 1044768-44-6
Molecular Formula C9H6ClFN2
Quantity 1 g
InChI Formula InChI=1S/C9H6ClFN2/c1-5-12-8-3-2-6(11)4-7(8)9(10)13-5/h2-4H,1H3
SMILES Cc1nc(Cl)c2cc(F)ccc2n1
IUPAC Name 4-chloro-6-fluoro-2-methylquinazoline
Molecular Weight (g/mol) 196.61
Formula Weight 196.0204