Learn More
4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone
Product Code. 11352326
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352326 5 g 5g
11362326 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11352326 Supplier Thermo Scientific Alfa Aesar Supplier No. L06011.06
Request Bulk or Custom Format
CAS: 37074-39-8 | C9H9ClO | 168.62 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37074-39-8
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045172
InChI Key XOIGZLJCLDWTQH-UHFFFAOYSA-N
Synonym 1-4-chloro-3-methylphenyl ethanone, 4'-chloro-3'-methylacetophenone, 4-chloro-3-methylacetophenone, ethanone, 1-4-chloro-3-methylphenyl, 1-4-chloro-3-methylphenyl ethan-1-one, 4-chloro-3-methyl acetophenone, 1-4-chloro-3-methyl-phenyl-ethanone, ksc577m5h, 3-methyl-4-chloroacetophenone, 2-methyl-4-acetyl-chlorbenzol
PubChem CID 592743
IUPAC Name 1-(4-chloro-3-methylphenyl)ethanone
SMILES CC(=O)C1=CC(C)=C(Cl)C=C1
Specifications
Density 1.15
Boiling Point 104°C to 105°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 774639
Formula Weight 168.62
Percent Purity 75%
Grade Technical
Assay remainder 3'-chloro-4'-methylacetophenone
Chemical Name or Material 4'-Chloro-3'-methylacetophenone