missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dimethylpicolinic Acid, TRC

Product Code. 30354793
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354793 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354793 Supplier Toronto Research Chemicals Supplier No. TRCC3668652.5G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-3,5-dimethylpicolinic Acid
CAS 447461-22-5
Molecular Formula C8H8ClNO2
Quantity 2.5 g
Synonym 4-Chloro-3,5-dimethyl-2-pyridinecarboxylic Acid
InChI Formula InChI=1S/C8H8ClNO2/c1-4-3-10-7(8(11)12)5(2)6(4)9/h3H,1-2H3,(H,11,12)
SMILES Cc1cnc(C(=O)O)c(C)c1Cl
IUPAC Name 4-chloro-3,5-dimethylpyridine-2-carboxylic acid
Molecular Weight (g/mol) 185.61
Formula Weight 185.0244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.