Learn More
4’-Chloro-2-hydroxy-benzophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 4'-Chloro-2-hydroxy-benzophenone |
| CAS | 2985-79-7 |
| Molecular Formula | C13H9ClO2 |
| Quantity | 50 mg |
| Synonym | 4-Chloro-2'-hydroxy-benzophenone, (4-chlorophenyl)(2-hydroxyphenyl)methanone |
| InChI Formula | InChI=1S/C13H9ClO2/c14-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)15/h1-8,15H |
| SMILES | ClC1=CC=C(C(C2=CC=CC=C2O)=O)C=C1 |
| IUPAC Name | (4-chlorophenyl)-(2-hydroxyphenyl)methanone |
| Molecular Weight (g/mol) | 232.66 |
| Formula Weight | 232.0291 |