Learn More
4-Carboxy Tolbutamide-d9 Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Carboxy Tolbutamide-d9 Methyl Ester |
| CAS | 1185126-97-9 |
| Molecular Formula | C13H9D9N2O5S |
| Quantity | 2.5 mg |
| Synonym | Methyl 4-Butyl-d9-aminocarbonylaminosulfonylbenzoate, 4-[[[(Butyl-d9-amino)carbonyl]amino]sulfonyl]-benzoic Acid Methyl Ester |
| InChI Formula | InChI=1S/C13H18N2O5S/c1-3-4-9-14-13(17)15-21(18,19)11-7-5-10(6-8-11)12(16)20-2/h5-8H,3-4,9H2,1-2H3,(H2,14,15,17)/i1D3,3D2,4D2,9D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC(=O)NS(=O)(=O)c1ccc(cc1)C(=O)OC |
| IUPAC Name | methyl 4-(1,1,2,2,3,3,4,4,4-nonadeuteriobutylcarbamoylsulfamoyl)benzoate |
| Molecular Weight (g/mol) | 323.41 |