Learn More
4-Bromophenylisothiocyanate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 1985-12-2 |
| Molecular Formula | C7H4BrNS |
| MDL Number | MFCD00004808 |
| Quantity | 25 g |
| SMILES | BrC1=CC=C(N=C=S)C=C1 |
| Molecular Weight (g/mol) | 214.08 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Bromophenylisothiocyanate |