Learn More
4-Bromophenetole, TRC
Product Code. 30335827
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335827 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30335827 Supplier Toronto Research Chemicals Supplier No. TRCB68615050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Bromophenetole
CAS 588-96-5
Molecular Formula C8H9BrO
Quantity 50 g
Synonym 1-Bromo-4-ethoxy-benzene, p-Bromo-phenetole, 1-Bromo-4-ethoxybenzene, 4-Bromoethoxybenzene, 4-Bromophenyl Ethyl Ether, 4-Ethoxybromobenzene, 4-Ethoxyphenyl Bromide, Ethyl 4-Bromophenyl Ether, NSC 8053, p-Bromoethoxybenzene, p-Bromophenetole, p-Bromophenol Ethyl Ether, p-Ethoxybromobenzene, p-Ethoxyphenyl Bromide
InChI Formula InChI=1S/C8H9BrO/c1-2-10-8-5-3-7(9)4-6-8/h3-6H,2H2,1H3
SMILES CCOc1ccc(Br)cc1
IUPAC Name 1-bromo-4-ethoxybenzene
Molecular Weight (g/mol) 201.06
Formula Weight 199.9837