missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobutyric-2,2,3,3,4,4-d6 acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Bromobutanoic Acid-D6 |
| CAS | 136091-75-3 |
| Molecular Formula | C4 D6 H Br O2 |
| Quantity | 0.1 g |
| Synonym | Butanoic-2,2,3,3,4,4-d6 acid, 4-bromo- (9 CI), 4-Bromobutanoic-2,2,3,3,4,4-d6 acid, 4-Bromobutanoic acid-D6, 4-Bromobutyric acid-d6, Carboxypropyl-d6 bromide, γ-Bromobutyric acid-d6 |
| InChI Formula | InChI=1 S/C4H7BrO2/c5-3-1-2-4(6)7/h1-3H2,(H,6,7)/i1D2,2D2,3D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 4-bromo-2,2,3,3,4,4-hexadeuteriobutanoic acid |
| Molecular Weight (g/mol) | 173.0382 |
| Formula Weight | 172.0006 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction