Learn More
4-Bromo-N,N-diethylaniline, 97%
Product Code. 11322246
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. 11322246
Brand: Thermo Scientific Alfa Aesar L03619.09
Request Bulk or Custom Format
CAS: 2052-06-4 | C10H14BrN | 228.13 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2052-06-4
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.13
MDL Number MFCD00013530
InChI Key NGYMZFJVHHKJQR-UHFFFAOYSA-N
PubChem CID 16328
IUPAC Name 4-bromo-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)Br
Specifications
Melting Point 32°C to 35°C
Boiling Point 269°C to 271°C
Quantity 10 g
UN Number UN2811
Beilstein 2803424
Sensitivity Light sensitive
Synonym p-bromo-n,n-diethylaniline, benzenamine, 4-bromo-n,n-diethyl, n,n-diethyl-p-bromoaniline, 4-n,n-diethylamino-bromobenzene, benzenamine,4-bromo-n,n-diethyl, p-n,nddiethylaniline, 4-bromodiethylaniline, 4-bromophenyldiethylamine, 4-bromophenyl diethylamine, 4-bromo-phenyl diethylamine
Solubility Information Insoluble in water.
Formula Weight 228.13
Percent Purity 97%
Chemical Name or Material 4-Bromo-N,N-diethylaniline