Learn More
4-Bromo-5-methylbenzene-1,3-diol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-5-methylbenzene-1,3-diol |
| CAS | 3446-04-6 |
| Molecular Formula | C7H7BrO2 |
| Quantity | 1 g |
| Synonym | 4-Bromo-5-methyl-1,3-benzenediol, 2-Bromo-3,5-dihydroxytoluene |
| InChI Formula | InChI=1S/C7H7BrO2/c1-4-2-5(9)3-6(10)7(4)8/h2-3,9-10H,1H3 |
| SMILES | Cc1cc(O)cc(O)c1Br |
| IUPAC Name | 4-bromo-5-methylbenzene-1,3-diol |
| Molecular Weight (g/mol) | 203.03 |
| Formula Weight | 201.9629 |