Learn More
4-Bromo-3-thiophenesulfonyl Chloride, Technical grade, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-3-thiophenesulfonyl Chloride, Technical grade |
| CAS | 111283-90-0 |
| Molecular Formula | C4 H2 Br Cl O2 S2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C4H2BrClO2S2/c5-3-1-9-2-4(3)10(6,7)8/h1-2H |
| SMILES | ClS(=O)(=O)c1cscc1Br |
| IUPAC Name | 4-bromothiophene-3-sulfonyl chloride |
| Molecular Weight (g/mol) | 261.54 |
| Formula Weight | 259.8368 |