Learn More
4-Bromo-3,5-dihydroxybenzamide, 97%, Thermo Scientific™
Product Code. 11385309
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11385309
Brand: Thermo Scientific Alfa Aesar B20493.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13429-12-4
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.03
MDL Number MFCD00017131
InChI Key GCNIEGHLVCGBRA-UHFFFAOYSA-N
Synonym 4-bromo-3,5-resorcylamide, benzamide, 4-bromo-3,5-dihydroxy, .alpha.-resorcylamide, 4-bromo, acmc-1bzb8, benzamide,4-bromo-3,5-dihydroxy, 4-bromanyl-3,5-bis oxidanyl benzamide, alpha-resorcylamide, 4-bromo-8ci
PubChem CID 83438
IUPAC Name 4-bromo-3,5-dihydroxybenzamide
SMILES NC(=O)C1=CC(O)=C(Br)C(O)=C1
Specifications
Quantity 10 g
Formula Weight 232.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-3,5-dihydroxybenzamide