Learn More
4-Bromo-3,5-difluorobenzoic acid, 96%, Thermo Scientific Chemicals
Product Code. 15496348
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496348 1 g 1g
15406358 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15496348 Supplier Thermo Scientific Alfa Aesar Supplier No. H59904.03
Request Bulk or Custom Format
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 651027-00-8
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237.00
MDL Number MFCD18917156
InChI Key NLLRAEQVOZOSBB-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluorobenzoicacid, 3,5-difluoro-4-bromobenzoic acid
PubChem CID 11424897
IUPAC Name 4-bromo-3,5-difluorobenzoic acid
SMILES OC(=O)C1=CC(F)=C(Br)C(F)=C1
Specifications
Melting Point 181°C to 185°C
Quantity 1 g
Beilstein 9620931
Formula Weight 237
Percent Purity 96%
Chemical Name or Material 4-Bromo-3,5-difluorobenzoic acid