missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 15411848
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15411848

Brand: Thermo Scientific Alfa Aesar H55545.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Bromo-2-methoxybenzaldehyde is used in the building block, synthesis of a nonacid degradable linker for solid-phase synthesis.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43192-33-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD08276823
InChI Key YMXQYZABFWVXEK-UHFFFAOYSA-N
PubChem CID 11053041
IUPAC Name 4-bromo-2-methoxybenzaldehyde
SMILES COC1=C(C=O)C=CC(Br)=C1

Specifications

Melting Point 67°C to 71°C
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 4-bromo-2-methoxy-benzaldehyde, benzaldehyde, 4-bromo-2-methoxy, 2-methoxy-4-bromobenzaldehyde, 4-bromo-2-methoxy benzaldehyde, acmc-209jte, 4-bromo-2-methoxybezaldehyde, ksc494s3l, 4-bromo-2-methoxybenz-aldehyde, benzaldehyde,4-bromo-2-methoxy, 4-bromanyl-2-methoxy-benzaldehyde
Solubility Information Soluble in water.
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methoxybenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.