Learn More
4-Bromo-2-fluorobenzaldehyde, 97%
Product Code. 11473147
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473147 100 g 100g
11453147 5 g 5g
11463147 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11473147 Supplier Thermo Scientific Alfa Aesar Supplier No. A16112.22
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
CAS: 57848-46-1 | C7H4BrFO | 203.01 g/mol
4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.
Solubility
Insoluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57848-46-1
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00143261
InChI Key UPCARQPLANFGQJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde
PubChem CID 143517
IUPAC Name 4-bromo-2-fluorobenzaldehyde
SMILES FC1=CC(Br)=CC=C1C=O
Specifications
Melting Point 59°C to 63°C
Boiling Point 42°C (19 mmHg)
Quantity 100 g
Beilstein 7700208
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 203.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluorobenzaldehyde