missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoroaniline, 98+%

Product Code. 11413923
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 11413923

Brand: Thermo Scientific Alfa Aesar A10228.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 367-24-8 | C6H5BrFN | 190.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-24-8
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00010221
InChI Key GZRMNMGWNKSANY-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromoaniline, benzenamine, 4-bromo-2-fluoro, 4-bromo-2-fluoro-phenylamine, 4-bromo-2-fluoro-aniline, 4-bromo-2-fluorobenzenamine, 4-bromo-2-fluoro aniline, 4-bromo-2-fluorophenylamine, 4-brom-2-fluoranilin, zlchem 545, pubchem1525
PubChem CID 123050
IUPAC Name 4-bromo-2-fluoroaniline
SMILES C1=CC(=C(C=C1Br)F)N

Specifications

Melting Point 38°C to 42°C
Boiling Point 103°C to 108°C (13 mmHg)
Flash Point 104°C (219°F)
Quantity 25 g
UN Number UN2811
Beilstein 2081089
Formula Weight 190.01
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-fluoroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.