missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-6-fluorophenol, 97+%, Thermo Scientific™

Catalog Number p-7027423
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
161045-79-0
C6H3BrClFO
225.44
MFCD01631476
QEYONPKSDTUPAX-UHFFFAOYSA-N
4-bromo-2-chloro-6-fluoro-phenol, phenol,4-bromo-2-chloro-6-fluoro, pubchem2841, acmc-209dlz, intermediates-zcf02212, 3-bromo-2-chloro-6-fluoro-phenol, phenol, 4-bromo-2-chloro-6-fluoro, 4-bromo-2-chloro-6-fluorophenol
2773270
4-bromo-2-chloro-6-fluorophenol
OC1=C(F)C=C(Br)C=C1Cl
This item is not returnable. View return policy
161045-79-0
C6H3BrClFO
225.44
MFCD01631476
QEYONPKSDTUPAX-UHFFFAOYSA-N
4-bromo-2-chloro-6-fluoro-phenol, phenol,4-bromo-2-chloro-6-fluoro, pubchem2841, acmc-209dlz, intermediates-zcf02212, 3-bromo-2-chloro-6-fluoro-phenol, phenol, 4-bromo-2-chloro-6-fluoro, 4-bromo-2-chloro-6-fluorophenol
2773270
4-bromo-2-chloro-6-fluorophenol
OC1=C(F)C=C(Br)C=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 255.45
Percent Purity ≥97%
Chemical Name or Material 4-Bromo-2-chloro-6-fluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'