missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dimethylanisole, 99%

Product Code. 11415027
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11415027

Brand: Thermo Scientific Alfa Aesar B21931.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14804-38-7 | C9H11BrO | 215.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14804-38-7
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00143259
InChI Key MMARFGDTMJBIBK-UHFFFAOYSA-N
Synonym 4-bromo-2,6-dimethylanisole, 5-bromo-2-methoxy-m-xylene, 4-brom-2,6-dimethylanisole, 1-bromo-3,5-dimethyl-4-methoxybenzene, 2,6-dimethyl-4-bromoanisole, benzene, 5-bromo-2-methoxy-1,3-dimethyl, timtec-bb sbb005776, attercop-chm at108629, 4-bromo-2,6-dimethyl-methoxybenzene, pubchem2654
PubChem CID 278654
IUPAC Name 5-bromo-2-methoxy-1,3-dimethylbenzene
SMILES COC1=C(C)C=C(Br)C=C1C

Specifications

Density 1.347
Boiling Point 117°C to 120°C (13 mmHg)
Quantity 25 g
Refractive Index 1.545
Beilstein 1940290
Formula Weight 215.09
Percent Purity 99%
Chemical Name or Material 4-Bromo-2,6-dimethylanisole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.