Learn More
4-Bromo-1,2-dimethoxybenzene, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-1,2-dimethoxybenzene |
| CAS | 2859-78-1 |
| Molecular Formula | C8H9BrO2 |
| Quantity | 100 g |
| Synonym | 4-Bromo-veratrole, 1,2-Dimethoxy-4-bromobenzene, 1-Bromo-3,4-dimethoxybenzene, 3,4-Dimethoxy-1-bromobenzene, 3,4-Dimethoxybromobenzene, 3,4-Dimethoxyphenyl bromide, 4-Bromo-1,2-dimethoxybenzene, 4-Bromo-2-methoxyanisole, 4-Bromoveratrole, Bromoveratrole, NSC 36284 |
| InChI Formula | InChI=1S/C8H9BrO2/c1-10-7-4-3-6(9)5-8(7)11-2/h3-5H,1-2H3 |
| SMILES | COc1ccc(Br)cc1OC |
| IUPAC Name | 4-bromo-1,2-dimethoxybenzene |
| Molecular Weight (g/mol) | 217.06 |
| Formula Weight | 215.9786 |