missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,1,2-trifluoro-1-butene, 98%

Product Code. 11303366
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11303366 50 g 50g
11393356 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 11303366

Brand: Thermo Scientific Alfa Aesar L12766.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10493-44-4 | C4H4BrF3 | 188.975 g/mol

4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10493-44-4
Molecular Formula C4H4BrF3
Molecular Weight (g/mol) 188.975
MDL Number MFCD00039274
InChI Key GQCQMFYIFUDARF-UHFFFAOYSA-N
Synonym 4-bromo-1,1,2-trifluoro-1-butene, 4-bromo-1,1,2-trifluorobutene-1, 1-butene, 4-bromo-1,1,2-trifluoro, 1,1,2-trifluoro-4-bromobutene, acmc-20amo8, timtec-bb sbb006604, gqcqmfyifudarf-uhfffaoysa, 4-bromo-1,1,2-trifluorobutene., 3,4,4-trifluoro-3-butenyl bromide, 3,4,4-trifluorobut-3-enyl bromide
PubChem CID 66333
IUPAC Name 4-bromo-1,1,2-trifluorobut-1-ene
SMILES C(CBr)C(=C(F)F)F

Specifications

Density 1.639
Boiling Point 95°C to 98°C
Flash Point 17°C (62°F)
Odor Recognizable
Linear Formula BrCH2CH2CF=CF2
Refractive Index 1.402
Quantity 50 g
UN Number UN1993
Beilstein 1840755
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 188.98
Percent Purity 98%
Chemical Name or Material 4-Bromo-1,1,2-trifluoro-1-butene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.