missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-aminomethyl)pyridine, 97%, Thermo Scientific™

Product Code. 15484878
Change view
Click to view available options
Quantity:
5 g
30 g
Unit Size:
30g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484878 30 g 30g
15474878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484878 Supplier Thermo Scientific Alfa Aesar Supplier No. H58647.15

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111080-65-0
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD07369734
InChI Key KXCFJDZVQLRYSI-UHFFFAOYSA-N
Synonym tert-butyl pyridin-4-ylmethyl carbamate, n-boc-4-aminomethylpyridine, 4-tert-butoxycarbonylamino methyl pyridine, tert-butyl n-pyridin-4-ylmethyl carbamate, pyridin-4-ylmethyl carbamic acid tert-butyl ester, tert-butyl pyridin-4-ylmethylcarbamate, 4-t-butyloxycarbonylaminomethyl-pyridine, 4-tert-butyloxycarbonylaminomethyl pyridin, 4-tert-butyloxycarbonylaminomethyl pyridine
PubChem CID 1512532
IUPAC Name tert-butyl N-(pyridin-4-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=NC=C1

Specifications

Melting Point 78°C to 85°C
Quantity 30 g
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 4-(Boc-aminomethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.