Learn More
4-Bis(2-cyano-3-pyridylmethyl)aminobenzonitrile, 97%, Thermo Scientific™
Product Code. 15474306
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15474306
Brand: Thermo Scientific Alfa Aesar H58503.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1421261-92-8
Molecular Formula C21H14N6
Molecular Weight (g/mol) 350.385
MDL Number MFCD22682981
InChI Key FPDANHUWWTXQDQ-UHFFFAOYSA-N
Synonym 3-4-cyanophenyl 2-cyanopyridin-3-yl methyl amino methyl pyridine-2-carbonitrile, 4-bis 2-cyano-3-pyridylmethyl aminobenzonitrile
PubChem CID 97030352
IUPAC Name 3-[[4-cyano-N-[(2-cyanopyridin-3-yl)methyl]anilino]methyl]pyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)CN(CC2=C(N=CC=C2)C#N)C3=CC=C(C=C3)C#N
Specifications
Quantity 5 g
UN Number UN3439
Formula Weight 350.38
Percent Purity 97%
Chemical Name or Material 4-Bis(2-cyano-3-pyridylmethyl)aminobenzonitrile