missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylyl Glucuronide, TRC

Product Code. 30335874
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335874 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335874 Supplier Toronto Research Chemicals Supplier No. TRCB39790525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Biphenylyl Glucuronide
CAS 19132-91-3
Molecular Formula C18 H18 O7
Quantity 25 mg
Synonym 4-Biphenylyl Glucopyranosiduronic Acid, β-D-4-Biphenylyl Glucopyranosiduronic Acid, 4-Glucuronosidobiphenyl, 4-Hydroxybiphenyl Glucuronide
InChI Formula InChI=1S/C18H18O7/c19-13-14(20)16(17(22)23)25-18(15(13)21)24-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9,13-16,18-21H,(H,22,23)/t13-,14-,15+,16-,18?/m0/s1
SMILES O[C@H]1C(Oc2ccc(cc2)c3ccccc3)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O
IUPAC Name (2S,3S,4S,5R)-3,4,5-trihydroxy-6-(4-phenylphenoxy)oxane-2-carboxylic acid
Molecular Weight (g/mol) 346.331
Formula Weight 346.105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.