missing translation for 'onlineSavingsMsg'
Learn More

4-[beta-(5-Methylpyrazinyl-2-carboxamido)ethyl]benzene Sulfonamide, TRC

Product Code. 30180355
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180355 500 mg 500mg
30180157 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180355 Supplier Toronto Research Chemicals Supplier No. TRCM325946500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33288-71-0
Molecular Formula C14 H16 N4 O3 S
Molecular Weight (g/mol) 320.37
Synonym 2-Pyrazinecarboxamide, N-[2-[4-(aminosulfonyl)phenyl]ethyl]-5-methyl-, Pyrazinecarboxamide, 5-methyl-N-(p-sulfamoylphenethyl)- (8CI), Pyrazinecarboxamide, N-[2-[4-(aminosulfonyl)phenyl]ethyl]-5-methyl- (9CI), N-[2-[4-(Aminosulfonyl)phenyl]ethyl]-5-methyl-2-pyrazinecarboxamide, 4-[2-(5-Methylpyrazine-2-carboxamido)ethyl]benzenesulfonamide, 4-[2-(5-Methylpyrazinyl-2-carboxamido)ethyl]benzenesulfonamide, 5-Methyl-N-[2-(4-sulfamoylphenyl)ethyl]pyrazine-2-carboxamide, 5-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide, 4-[β-(5-Methylpyrazinyl-2-carboxamido)ethyl]benzene Sulfonamide, N-{2-[(4-Aminosulfonyl)phenyl]ethyl}-5-methylpyrazinecarboxamide
IUPAC Name 5-methyl-N-[2-(4-sulfamoylphenyl)ethyl]pyrazine-2-carboxamide
SMILES Cc1cnc(cn1)C(=O)NCCc2ccc(cc2)S(=O)(=O)N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H16N4O3S/c1-10-8-18-13(9-17-10)14(19)16-7-6-11-2-4-12(5-3-11)22(15,20)21/h2-5,8-9H,6-7H2,1H3,(H,16,19)(H2,15,20,21)
Formula Weight 320.0943
Chemical Name or Material 5-Methyl-N-[2-(4-sulfamoylphenyl)ethyl]pyrazine-2-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.