missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxymethyl-2-(4-methoxyphenyl)-4-methyl-d3-1,3-dioxolane-5-carboxylic Acid Methyl Ester, TRC

Product Code. 30336154
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336154 1 mg 1mg
30346519 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30336154 Supplier Toronto Research Chemicals Supplier No. TRCB2866521MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Benzyloxymethyl-2-(4-methoxyphenyl)-4-methyl-d3-1,3-dioxolane-5-carboxylic Acid Methyl Ester
Molecular Formula C21 D3 H21 O6
Quantity 1 mg
InChI Formula InChI=1S/C21H24O6/c1-21(14-25-13-15-7-5-4-6-8-15)18(19(22)24-3)26-20(27-21)16-9-11-17(23-2)12-10-16/h4-12,18,20H,13-14H2,1-3H3/t18-,20?,21+/m1/s1/i1D3
SMILES [2H]C([2H])([2H])[C@@]1(COCc2ccccc2)OC(O[C@@H]1C(=O)OC)c3ccc(OC)cc3
IUPAC Name methyl (4S,5S)-2-(4-methoxyphenyl)-5-(phenylmethoxymethyl)-5-(trideuteriomethyl)-1,3-dioxolane-4-carboxylate
Molecular Weight (g/mol) 375.43
Formula Weight 375.176
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.