missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)cyclohexanol (cis / trans mixture), TRC

Product Code. 30340125
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340125 Supplier Toronto Research Chemicals Supplier No. TRCB2863905G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Benzyloxy)cyclohexanol
CAS 2976-80-9
Molecular Formula C13 H18 O2
Quantity 5 g
Synonym 4-(Benzyloxy)cyclohexanol (cis / trans mixture)
InChI Formula InChI=1S/C13H18O2/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11/h1-5,12-14H,6-10H2
SMILES OC1CCC(CC1)OCc2ccccc2
IUPAC Name 4-phenylmethoxycyclohexan-1-ol
Molecular Weight (g/mol) 206.28
Formula Weight 206.1307
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.