missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3,5-dimethylbenzaldehyde, 95%

Product Code. 11409577
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11409577

Brand: Thermo Scientific Alfa Aesar B22067.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144896-51-5 | C16H16O2 | 240.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144896-51-5
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.30
MDL Number MFCD00800683
InChI Key GSYUTKRSEZMBNC-UHFFFAOYSA-N
Synonym 4-benzyloxy-3,5-dimethylbenzaldehyde, 3,5-dimethyl-4-phenylmethoxy benzaldehyde, zlchem 1072, acmc-1bxy6, 3,5-dimethyl-4-benzyloxybenzaldehyde, 3,5-dimethyl-4-benzyloxy benzaldehyde, 4-benzyloxy-3,5-dimethyl-benzaldehyde
PubChem CID 563557
IUPAC Name 3,5-dimethyl-4-phenylmethoxybenzaldehyde
SMILES CC1=CC(C=O)=CC(C)=C1OCC1=CC=CC=C1

Specifications

Density 1.1
Boiling Point 162°C to 163°C (1 mmHg)
Quantity 5 g
Refractive Index 1.586
Formula Weight 240.3
Percent Purity 95%
Chemical Name or Material 4-Benzyloxy-3,5-dimethylbenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.