Learn More
4-Aminopyrene, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Aminopyrene |
| CAS | 17075-03-5 |
| Molecular Formula | C16H11N |
| Quantity | 50 mg |
| Synonym | Pyren-4-amine |
| InChI Formula | InChI=1S/C16H11N/c17-14-9-12-5-1-3-10-7-8-11-4-2-6-13(14)16(11)15(10)12/h1-9H,17H2 |
| SMILES | Nc1cc2cccc3ccc4cccc1c4c23 |
| IUPAC Name | pyren-4-amine |
| Molecular Weight (g/mol) | 217.27 |
| Formula Weight | 217.0891 |