Learn More
4-Aminobiphenyl-d9, CDN
Product Code. 30319887
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319887
Brand: CDN CDND26380.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Aminobiphenyl D9
CAS 344298-96-0
Molecular Formula C12 2H9 H2 N
Quantity 0.05 g
Synonym [1,1'-Biphenyl-2,2',3,3',4',5,5',6,6'-d9]-4-amine (9 CI), 4-Aminobiphenyl D9, 4-Aminobiphenyl-D9
InChI Formula InChI=1 S/C12H11N/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9 H,13H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D
SMILES [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c([2 H])c(N)c([2 H])c2[2 H]
IUPAC Name 2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)aniline
Molecular Weight (g/mol) 178.28
Formula Weight 178.1456 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity