Learn More
4-Amino-N-(2-ethylphenyl)benzamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-N-(2-ethylphenyl)benzamide |
| CAS | 29027-73-4 |
| Molecular Formula | C15H16N2O |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C15H16N2O/c1-2-11-5-3-4-6-14(11)17-15(18)12-7-9-13(16)10-8-12/h3-10H,2,16H2,1H3,(H,17,18) |
| SMILES | CCc1ccccc1NC(=O)c2ccc(N)cc2 |
| IUPAC Name | 4-amino-N-(2-ethylphenyl)benzamide |
| Molecular Weight (g/mol) | 240.3 |
| Formula Weight | 240.1263 |