Learn More
4-Amino-3,5-dichlorodifluoropyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-3,5-dichlorodifluoropyridine |
| CAS | 2840-00-8 |
| Molecular Formula | C5H2Cl2F2N2 |
| Quantity | 10 g |
| Synonym | 3,5-Dichloro-2,6-difluoro-4-pyridinamine, (3,5-Dichloro-2,6-difluoropyridin-4-yl)amine, 2,6-Difluoro-3,5-dichloro-4-aminopyridine, 3,5-Dichloro-2,6-difluoropyrid-4-ylamine, 4-Amino-3,5-dichloro-2,6-difluoropyridine |
| InChI Formula | InChI=1S/C5H2Cl2F2N2/c6-1-3(10)2(7)5(9)11-4(1)8/h(H2,10,11) |
| SMILES | Nc1c(Cl)c(F)nc(F)c1Cl |
| IUPAC Name | 3,5-dichloro-2,6-difluoropyridin-4-amine |
| Molecular Weight (g/mol) | 198.99 |
| Formula Weight | 197.9563 |