Learn More
4-Amino-2-methyl-5-pyrimidinemethanesulfonic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-2-methyl-5-pyrimidinemethanesulfonic acid |
| CAS | 2908-73-8 |
| Molecular Formula | C6H9N3O3S |
| Quantity | 250 mg |
| Synonym | 2-Methyl-4-amino-5-sulfomethylpyrimidine, NSC 165509 |
| InChI Formula | InChI=1S/C6H9N3O3S/c1-4-8-2-5(6(7)9-4)3-13(10,11)12/h2H,3H2,1H3,(H2,7,8,9)(H,10,11,12) |
| SMILES | Cc1ncc(CS(=O)(=O)O)c(N)n1 |
| IUPAC Name | (4-amino-2-methylpyrimidin-5-yl)methanesulfonic acid |
| Molecular Weight (g/mol) | 203.22 |
| Formula Weight | 203.0365 |