Learn More
4-Allyl-3-Thiosemicarbazide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Allyl-3-Thiosemicarbazide |
| CAS | 3766-55-0 |
| Molecular Formula | C4H9N3S |
| Quantity | 1 g |
| Synonym | Hydrazinecarbothioamide, N-2-propenyl- (9CI), Semicarbazide, 4-allyl-3-thio- (6CI,7CI,8CI), N-2-Propen-1-ylhydrazinecarbothioamide, 1-Amino-3-(prop-2-en-1-yl)thiourea, 1-Amino-3-prop-2-enylthiourea, 4-(2-Propenyl)hydrazinethiocarbamide, 4-Allyl-3-thiosemicarbazide4-Allylthiosemicarbazide, 4-Propenylthiosemicarbazide, Allylthiosemicarbazide, N-(2-Propenyl)hydrazinecarbothioamide, N-Allylthiosemicarbazide, NSC 75830 |
| InChI Formula | InChI=1S/C4H9N3S/c1-2-3-6-4(8)7-5/h2H,1,3,5H2,(H2,6,7,8) |
| SMILES | NNC(=S)NCC=C |
| IUPAC Name | 1-amino-3-prop-2-enylthiourea |
| Molecular Weight (g/mol) | 131.2 |
| Formula Weight | 131.0517 |