missing translation for 'onlineSavingsMsg'
Learn More

4,8-Dimethyl-1,3(E),7-nonatriene, TRC

Product Code. 30380172
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380172 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380172 Supplier Toronto Research Chemicals Supplier No. TRCD47581025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,8-Dimethyl-1,3(E),7-nonatriene
CAS 19945-61-0
Purity Grade Notes GC
Molecular Formula C11 H18
Quantity 25 mg
Synonym (E)-4,8-Dimethyl-1,3,7-nonatriene
InChI Formula InChI=1S/C11H18/c1-5-7-11(4)9-6-8-10(2)3/h5,7-8H,1,6,9H2,2-4H3/b11-7+
SMILES CC(=CCC\C(=C\C=C)\C)C
IUPAC Name (3E)-4,8-dimethylnona-1,3,7-triene
Molecular Weight (g/mol) 150.261
Formula Weight 150.1409
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.